Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261050.1 | NP_001247979.1 | 173 | 94 | ||
XM_015122785.2 | XP_014978271.1 | 211 | 94 | |||
GenBank mRNA | JU471006.1 | - | 173 | 94 | ||
JV601589.1 | - | 147 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |