Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC114675313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842554.1 | XP_028698387.1 | 483 | 93 |
Gene Symbol: CNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261809.1 | NP_001248738.1 | 730 | 93 | ||
XM_015145070.2 | XP_015000556.1 | 569 | 93 | |||
GenBank mRNA | JU319626.1 | - | 633 | 93 | ||
JU472900.1 | - | 673 | 93 | |||
JV044008.1 | - | 730 | 93 | |||
JV451657.1 | - | 748 | 93 |