Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266802.1 | NP_001253731.1 | 738 | 146 | ||
XM_028843445.1 | XP_028699278.1 | 848 | 146 | |||
GenBank mRNA | JV046696.1 | - | 738 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |