Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266544.1 | NP_001253473.1 | 351 | 86 | ||
GenBank mRNA | JU320285.1 | - | 338 | 86 | ||
JU471158.1 | - | 351 | 86 | |||
JV044116.1 | - | 351 | 86 | |||
JV518108.1 | - | 300 | 86 |