Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258176.1 | NP_001245105.1 | 1037 | 87 | ||
GenBank mRNA | JU335497.1 | - | 1037 | 87 | ||
JV622526.1 | - | 1108 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |