Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194131.2 | NP_001181060.1 | 477 | 94 | ||
XM_015143273.2 | XP_014998759.1 | 515 | 94 | |||
GenBank mRNA | JU320260.1 | - | 456 | 94 | ||
JU473207.1 | - | 476 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |