Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097639.4 | XP_001097639.2 | 105 | 104 | ||
GenBank mRNA | JU485288.1 | - | 83 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |