Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BROX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099998.4 | XP_001099998.4 | 734 | 121 | ||
XM_001100089.4 | XP_001100089.1 | 711 | 121 | |||
XM_028853490.1 | XP_028709323.1 | 622 | 121 | |||
XM_028853495.1 | XP_028709328.1 | 446 | 121 | |||
GenBank mRNA | JU474408.1 | - | 598 | 121 | ||
JV643219.1 | - | 648 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |