Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD1LG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083599.2 | NP_001077068.1 | 162 | 145 | ||
XM_015117664.2 | XP_014973150.1 | 385 | 145 | |||
GenBank mRNA | EF444815.1 | - | 163 | 145 | ||
EF444817.1 | - | 104 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |