Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266533.2 | NP_001253462.1 | 307 | 109 | ||
XM_015131545.2 | XP_014987031.2 | 184 | 109 | |||
XM_015131546.2 | XP_014987032.1 | 398 | 109 | |||
GenBank mRNA | JU325229.1 | - | 239 | 109 | ||
JU473772.1 | - | 292 | 109 | |||
JV636173.1 | - | 309 | 109 | |||
JV715754.1 | - | 261 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |