Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BUD31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194526.2 | NP_001181455.1 | 523 | 65 | ||
XM_002803209.3 | XP_002803255.1 | 412 | 65 | |||
XM_015134325.2 | XP_014989811.1 | 620 | 65 | |||
GenBank mRNA | JV601400.1 | - | 466 | 65 |
Gene Symbol: LOC718712 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV647815.1 | - | 3097 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |