Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257688.1 | NP_001244617.1 | 235 | 91 | ||
XM_015135934.2 | XP_014991420.1 | 464 | 91 | |||
GenBank mRNA | HE864465.1 | - | 189 | 91 | ||
HE864471.1 | - | 189 | 91 | |||
HE864472.1 | - | 189 | 91 | |||
JU320237.1 | - | 235 | 91 | |||
JU470729.1 | - | 307 | 91 | |||
JV046252.1 | - | 321 | 91 | |||
JV528445.1 | - | 319 | 91 | |||
JV639449.1 | - | 305 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |