Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135797.1 | NP_001129269.1 | 465 | 67 | ||
XM_015124402.2 | XP_014979888.2 | 734 | 67 | |||
XM_015124404.2 | XP_014979890.2 | 730 | 67 | |||
XM_015124405.2 | XP_014979891.2 | 649 | 67 | |||
XM_015124406.2 | XP_014979892.2 | 475 | 67 | |||
XM_015124407.2 | XP_014979893.2 | 740 | 67 | |||
XM_028838507.1 | XP_028694340.1 | 755 | 67 | |||
XM_028838508.1 | XP_028694341.1 | 721 | 67 | |||
XM_028838509.1 | XP_028694342.1 | 736 | 67 | |||
XM_028838510.1 | XP_028694343.1 | 752 | 67 | |||
XM_028838511.1 | XP_028694344.1 | 773 | 67 | |||
XM_028838512.1 | XP_028694345.1 | 769 | 67 | |||
XM_028838513.1 | XP_028694346.1 | 339 | 67 | |||
XM_028838514.1 | XP_028694347.1 | 755 | 67 | |||
XM_028838515.1 | XP_028694348.1 | 668 | 67 | |||
XM_028838516.1 | XP_028694349.1 | 668 | 67 | |||
GenBank mRNA | EU204919.1 | - | 465 | 67 | ||
JV600278.1 | - | 635 | 67 | |||
JV650561.1 | - | 672 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |