Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001117975.4 | XP_001117975.1 | 173 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |