Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194667.2 | NP_001181596.1 | 364 | 112 | ||
GenBank mRNA | JU335968.1 | - | 370 | 112 | ||
JV047964.1 | - | 370 | 112 | |||
JV659114.1 | - | 397 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |