Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159300.2 | NP_001152772.1 | 478 | 78 | ||
GenBank mRNA | CK231700.1 | - | 279 | 78 | ||
JV046823.1 | - | 355 | 78 | |||
JV649197.1 | - | 298 | 78 | |||
JV712955.1 | - | 344 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |