Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257479.1 | NP_001244408.1 | 520 | 149 | ||
XM_015120908.2 | XP_014976394.1 | 519 | 149 | |||
XM_028837139.1 | XP_028692972.1 | 898 | 149 | |||
GenBank mRNA | JU333358.1 | - | 520 | 149 | ||
JU474655.1 | - | 505 | 149 | |||
JV046431.1 | - | 519 | 149 | |||
JV459962.1 | - | 468 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |