Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANP32A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260818.1 | NP_001247747.1 | 351 | 143 | ||
GenBank mRNA | JV048333.1 | - | 351 | 143 | ||
JV521688.1 | - | 299 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |