Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164428.1 | NP_001157900.1 | 104 | 135 | ||
GenBank mRNA | GQ205391.1 | - | 104 | 135 | ||
JU471758.1 | - | 223 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta Inventoried |