Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258104.1 | NP_001245033.1 | 83 | 98 | ||
GenBank mRNA | JU473808.1 | - | 83 | 98 | ||
JV046906.1 | - | 109 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |