Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257722.1 | NP_001244651.1 | 221 | 79 | ||
XM_015127819.2 | XP_014983305.1 | 582 | 79 | |||
XM_015127820.2 | XP_014983306.1 | 416 | 79 | |||
XM_015127821.2 | XP_014983307.1 | 388 | 79 | |||
GenBank mRNA | JU321866.1 | - | 249 | 79 | ||
JU321867.1 | - | 221 | 79 | |||
JU472073.1 | - | 215 | 79 | |||
JV043803.1 | - | 238 | 79 | |||
JV043804.1 | - | 210 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |