Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARGLU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194022.1 | NP_001180951.1 | 588 | 82 | ||
GenBank mRNA | JU335222.1 | - | 447 | 82 | ||
JU335223.1 | - | 447 | 82 | |||
JU476185.1 | - | 458 | 82 | |||
JU476186.1 | - | 458 | 82 | |||
JU476187.1 | - | 458 | 82 | |||
JV047495.1 | - | 524 | 82 | |||
JV585921.1 | - | 430 | 82 | |||
JV636510.1 | - | 768 | 82 | |||
JV688400.1 | - | 577 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |