Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU335295.1 | - | 91 | 142 | ||
JU476253.1 | - | 95 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |