Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126364.2 | XP_014981850.1 | 435 | 96 | ||
GenBank mRNA | JU328877.1 | - | 90 | 96 | ||
JU489271.1 | - | 129 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |