Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5AR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001112923.4 | XP_001112923.1 | 1341 | 87 | ||
XM_001112956.4 | XP_001112956.1 | 1576 | 87 | |||
XM_015124346.2 | XP_014979832.1 | 1009 | 87 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta |