Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105717.4 | XP_001105717.2 | 1930 | 134 | ||
GenBank mRNA | JU476483.1 | - | 1851 | 134 | ||
JV066251.1 | - | 139 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |