Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144778.2 | XP_015000264.1 | 908 | 99 | ||
XM_028852362.1 | XP_028708195.1 | 893 | 99 | |||
XM_028852363.1 | XP_028708196.1 | 351 | 99 | |||
GenBank mRNA | JV514829.1 | - | 377 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |