Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLYWCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015125465.2 | XP_014980951.2 | 1920 | 106 | ||
XM_015125466.2 | XP_014980952.2 | 2310 | 106 | |||
XM_015125467.2 | XP_014980953.1 | 2056 | 106 | |||
XM_028841445.1 | XP_028697278.1 | 2766 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |