Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113643.4 | XP_001113643.1 | 671 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |