Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001106627.4 | XP_001106627.3 | 899 | 111 | ||
XM_028835123.1 | XP_028690956.1 | 899 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |