Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266356.1 | NP_001253285.1 | 1061 | 147 | ||
XM_015140114.2 | XP_014995600.1 | 1217 | 147 | |||
XM_015140115.2 | XP_014995601.1 | 1505 | 147 | |||
XM_015140116.2 | XP_014995602.1 | 1199 | 147 | |||
XM_015140117.2 | XP_014995603.1 | 944 | 147 | |||
XM_015140118.2 | XP_014995604.1 | 985 | 147 | |||
GenBank mRNA | JU336462.1 | - | 1213 | 147 | ||
JU336463.1 | - | 1195 | 147 | |||
JU336464.1 | - | 1195 | 147 | |||
JU336465.1 | - | 1195 | 147 | |||
JU336466.1 | - | 1195 | 147 | |||
JU472539.1 | - | 1174 | 147 | |||
JU472540.1 | - | 1192 | 147 | |||
JV044090.1 | - | 1061 | 147 | |||
JV044091.1 | - | 1043 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |