Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB33B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257437.2 | NP_001244366.1 | 311 | 102 | ||
XM_015139219.2 | XP_014994705.1 | 345 | 102 | |||
GenBank mRNA | JU336599.1 | - | 311 | 102 | ||
JU336600.1 | - | 311 | 102 | |||
JU477418.1 | - | 410 | 102 | |||
JV072784.1 | - | 446 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |