Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261472.1 | NP_001248401.1 | 373 | 149 | ||
GenBank mRNA | JU336606.1 | - | 373 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |