Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266682.1 | NP_001253611.1 | 1177 | 152 | ||
XM_015146191.2 | XP_015001677.1 | 1269 | 152 | |||
XM_015146253.2 | XP_015001739.2 | 1032 | 152 | |||
XM_015146255.2 | XP_015001741.2 | 1032 | 152 | |||
XM_015146257.2 | XP_015001743.2 | 1032 | 152 | |||
XM_028852160.1 | XP_028707993.1 | 1032 | 152 | |||
XM_028852161.1 | XP_028707994.1 | 1032 | 152 | |||
XM_028852162.1 | XP_028707995.1 | 1032 | 152 | |||
XM_028852164.1 | XP_028707997.1 | 1032 | 152 | |||
XM_028852165.1 | XP_028707998.1 | 1490 | 152 | |||
GenBank mRNA | JU336679.1 | - | 1156 | 152 | ||
JU336680.1 | - | 1152 | 152 | |||
JU477476.1 | - | 1229 | 152 | |||
JU477477.1 | - | 1225 | 152 | |||
JV048427.1 | - | 1181 | 152 | |||
JV048428.1 | - | 1177 | 152 | |||
JV630765.1 | - | 1251 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |