Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015143861.2 | XP_014999347.2 | 1691 | 92 | ||
XM_015143862.2 | XP_014999348.2 | 2001 | 92 | |||
XM_015143863.2 | XP_014999349.2 | 660 | 92 | |||
XM_015143864.2 | XP_014999350.2 | 1499 | 92 | |||
XM_015143865.2 | XP_014999351.2 | 1776 | 92 | |||
XM_015143866.2 | XP_014999352.2 | 1776 | 92 | |||
XM_015143867.2 | XP_014999353.2 | 341 | 92 | |||
XM_028851599.1 | XP_028707432.1 | 1776 | 92 | |||
XM_028851600.1 | XP_028707433.1 | 457 | 92 | |||
GenBank mRNA | JU323135.1 | - | 403 | 92 | ||
JU473407.1 | - | 436 | 92 | |||
JV046084.1 | - | 367 | 92 | |||
JV638901.1 | - | 394 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |