Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084680.4 | XP_001084680.1 | 259 | 98 | ||
XM_001084801.3 | XP_001084801.1 | 474 | 98 | |||
XM_028851555.1 | XP_028707388.1 | 633 | 98 | |||
XM_028851556.1 | XP_028707389.1 | 196 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |