Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDGFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099743.4 | XP_001099743.1 | 370 | 96 | ||
GenBank mRNA | JV635977.1 | - | 247 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |