Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H4orf36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190949.1 | NP_001177878.1 | 64 | 118 | ||
XM_015138740.2 | XP_014994226.1 | 342 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |