Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDR42E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194527.3 | NP_001181456.1 | 161 | 125 | ||
XM_015126670.2 | XP_014982156.1 | 464 | 125 | |||
XM_015126671.2 | XP_014982157.1 | 162 | 125 | |||
GenBank mRNA | JU323167.1 | - | 102 | 125 | ||
JV637606.1 | - | 116 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |