Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115919.2 | XP_014971405.1 | 233 | 139 | ||
XM_015115920.2 | XP_014971406.1 | 435 | 139 | |||
XM_015115922.2 | XP_014971408.1 | 233 | 139 | |||
XM_028832727.1 | XP_028688560.1 | 671 | 139 | |||
XM_028832728.1 | XP_028688561.1 | 628 | 139 | |||
GenBank mRNA | JU332796.1 | - | 139 | |||
JU474211.1 | - | 138 | 139 | |||
JU474212.1 | - | 138 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |