Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001114166.4 | XP_001114166.1 | 374 | 143 | ||
XM_015140279.2 | XP_014995765.1 | 728 | 143 | |||
XM_015140289.2 | XP_014995775.1 | 730 | 143 | |||
XM_015140310.2 | XP_014995796.1 | 708 | 143 | |||
XM_028833456.1 | XP_028689289.1 | 730 | 143 | |||
GenBank mRNA | JU323836.1 | - | 333 | 143 | ||
JU475074.1 | - | 299 | 143 | |||
JV711576.1 | - | 290 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |