Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194293.4 | NP_001181222.2 | 321 | 149 | ||
XM_015125596.2 | XP_014981082.2 | 314 | 149 | |||
GenBank mRNA | JV641670.1 | - | 151 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |