Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136113.2 | XP_014991599.1 | 2095 | 95 | ||
XM_015136114.2 | XP_014991600.1 | 2027 | 95 | |||
GenBank mRNA | JU474693.1 | - | 1437 | 95 | ||
JV046455.1 | - | 1528 | 95 | |||
JV636029.1 | - | 1609 | 95 | |||
JV709562.1 | - | 1429 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |