Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261511.1 | NP_001248440.1 | 420 | 95 | ||
GenBank mRNA | JU332830.1 | - | 420 | 95 | ||
JV494010.1 | - | 240 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |