Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148775.2 | XP_015004261.2 | 4087 | 71 | ||
GenBank mRNA | JV642262.1 | - | 212 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |