Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11H12orf61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194719.1 | NP_001181648.1 | 81 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |