Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2D6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040218.1 | NP_001035308.1 | 667 | 133 | ||
GenBank mRNA | AY635464.1 | - | 667 | 133 | ||
GU981763.1 | - | 675 | 133 | |||
JV708008.1 | - | 324 | 133 |
Gene Symbol: LOC100424965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150493.2 | XP_015005979.1 | 766 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |