Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266614.1 | NP_001253543.1 | 203 | 117 | ||
XM_028832684.1 | XP_028688517.1 | 753 | 117 | |||
GenBank mRNA | JU326174.1 | - | 186 | 117 | ||
JV635792.1 | - | 203 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |