Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028847575.1 | XP_028703408.1 | 1737 | 102 | ||
XM_028847576.1 | XP_028703409.1 | 1641 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |