Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266298.1 | NP_001253227.1 | 442 | 70 | ||
GenBank mRNA | JU334658.1 | - | 442 | 70 | ||
JV047160.1 | - | 366 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |